Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mterfd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199387.1 | NP_955419.1 | 1 - 2 | 104 | 85 | |
XM_006241509.2 | XP_006241571.1 | 1 - 2 | 130 | 85 | ||
GenBank mRNA | BC062080.1 | - | 1 - 2 | 104 | 5599618 | 85 |
FQ232616.1 | - | 1 - 2 | 117 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |