Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adck5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135798.1 | NP_001129270.1 | 13 - 14 | 1631 | 88 | |
XM_006241827.2 | XP_006241889.1 | 13 - 14 | 1535 | 88 | ||
GenBank mRNA | BC168693.1 | - | 13 - 14 | 1619 | 7935927 | 88 |
BP468309.1 | - | 2 - 3 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |