Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsta3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127455.2 | NP_001120927.1 | 1 - 2 | 33 | 89 | |
GenBank mRNA | BF289545.1 | - | 1 - 2 | 34 | 89 | |
FQ229919.1 | - | 1 - 2 | 44 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |