Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bai1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170597.2 | NP_001164068.2 | 9 - 10 | 2151 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |