Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134865.1 | NP_001128337.1 | 10 - 11 | 2519 | 100 | |
XM_008765548.1 | XP_008763770.1 | 10 - 11 | 2541 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |