Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241973.2 | XP_006242035.1 | 1 - 2 | 1500 | 79 | |
XM_008765679.1 | XP_008763901.1 | 2 - 3 | 1256 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |