Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241977.2 | XP_006242039.1 | 6 - 7 | 912 | 168 |
Set Membership: |
Amplicon length greater than or equal to 101 Ion Channel 3' Most Rattus norvegicus 1700 Probe spans exons |