Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpys | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031705.1 | NP_113893.1 | 5 - 6 | 1049 | 64 | |
GenBank mRNA | BC081768.1 | - | 5 - 6 | 1004 | 7107549 | 64 |
D63704.1 | - | 5 - 6 | 1049 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |