Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam91a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127578.1 | NP_001121050.1 | 5 - 6 | 657 | 98 | |
XM_006241684.2 | XP_006241746.1 | 5 - 6 | 661 | 98 | ||
GenBank mRNA | BC166852.1 | - | 5 - 6 | 657 | 9092514 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |