Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam91a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127578.1 | NP_001121050.1 | 9 - 10 | 1037 | 152 | |
XM_006241684.2 | XP_006241746.1 | 9 - 10 | 1041 | 152 | ||
GenBank mRNA | BC166852.1 | - | 9 - 10 | 1037 | 9092514 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |