Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Micall1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729832.4 | XP_002729878.2 | 11 - 12 | 2428 | 85 | |
XM_006242043.2 | XP_006242105.1 | 11 - 12 | 3047 | 85 | ||
XM_006242044.2 | XP_006242106.1 | 8 - 9 | 2151 | 85 | ||
XM_008765803.1 | XP_008764025.1 | 11 - 12 | 2630 | 85 | ||
XM_008765804.1 | XP_008764026.1 | 11 - 12 | 2762 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |