Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133313.2 | NP_579847.1 | 9 - 10 | 1268 | 99 | |
GenBank mRNA | AJ301634.1 | - | 9 - 10 | 1377 | 99 | |
BC062087.1 | - | 9 - 10 | 1268 | 5621521 | 99 | |
FQ213088.1 | - | 4 - 5 | 500 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |