Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf21b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130680.1 | NP_001124152.1 | 2 - 3 | 529 | 99 | |
XM_008765685.1 | XP_008763907.1 | 2 - 3 | 116 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |