Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130566.1 | NP_001124038.1 | 6 - 7 | 1512 | 73 | |
XM_008765580.1 | XP_008763802.1 | 5 - 6 | 3986 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Protein Phosphatase Detect Genomic DNA Rattus norvegicus Probe spans exons |