Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ribc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013949.1 | NP_001013971.2 | 2 - 3 | 371 | 122 | |
GenBank mRNA | BC079434.1 | - | 2 - 3 | 371 | 7115185 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |