Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naprt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207609.1 | NP_997492.1 | 5 - 6 | 769 | 130 | |
GenBank mRNA | AY214330.1 | - | 5 - 6 | 769 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |