Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrcc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139080.2 | NP_620780.2 | 2 - 3 | 154 | 101 | |
XM_008765671.1 | XP_008763893.1 | 2 - 3 | 147 | 101 | ||
GenBank mRNA | AB066102.1 | - | 1 - 2 | 138 | 101 | |
BC078718.1 | - | 2 - 3 | 154 | 7115290 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |