Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008378.1 | NP_001008379.1 | 2 - 3 | 327 | 85 | |
GenBank mRNA | BC086452.1 | - | 2 - 3 | 327 | 7307024 | 85 |
FQ214182.1 | - | 2 - 3 | 297 | 85 | ||
FQ218380.1 | - | 2 - 3 | 297 | 85 | ||
FQ219469.1 | - | 2 - 3 | 320 | 85 | ||
FQ219661.1 | - | 2 - 3 | 325 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |