Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phkg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080584.4 | NP_542151.1 | 8 - 9 | 1268 | 70 | |
XM_008759855.1 | XP_008758077.1 | 9 - 10 | 1091 | 70 | ||
XM_008759856.1 | XP_008758078.1 | 9 - 10 | 1209 | 70 | ||
GenBank mRNA | BC107906.1 | - | 8 - 9 | 1128 | 70 | |
BC128715.1 | - | 8 - 9 | 1197 | 7456955 | 70 | |
M73808.1 | - | 8 - 9 | 1268 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |