Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Micalcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182669.2 | NP_872610.2 | 2 - 3 | 138 | 70 | |
GenBank mRNA | BC098647.1 | - | 2 - 3 | 138 | 7112444 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |