Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bckdk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019244.2 | NP_062117.2 | 3 - 4 | 522 | 90 | |
GenBank mRNA | CK479351.1 | - | 3 - 4 | 522 | 90 | |
M93271.1 | - | 387 | 90 | |||
U27456.1 | - | 2 - 3 | 176 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |