Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031357.1 | NP_112647.1 | 8 - 9 | 1077 | 141 | |
GenBank mRNA | AB043870.1 | - | 8 - 9 | 1077 | 141 | |
AY724484.1 | - | 8 - 9 | 1078 | 141 | ||
BC081775.1 | - | 8 - 9 | 1086 | 7107183 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |