Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017464.2 | NP_001017464.1 | 5 - 6 | 630 | 133 | |
XM_006230403.2 | XP_006230465.1 | 5 - 6 | 730 | 133 | ||
XM_008759903.1 | XP_008758125.1 | 5 - 6 | 730 | 133 | ||
XM_008759904.1 | XP_008758126.1 | 3 - 4 | 396 | 133 | ||
XM_008759905.1 | XP_008758127.1 | 5 - 6 | 730 | 133 | ||
XM_008759906.1 | XP_008758128.1 | 5 - 6 | 730 | 133 | ||
GenBank mRNA | BC079317.1 | - | 5 - 6 | 633 | 7135047 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |