Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759812.1 | XP_008758034.1 | 6 - 7 | 1697 | 74 | |
GenBank mRNA | AF136401.1 | - | 1704 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |