Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009645.1 | NP_001009645.1 | 10 - 11 | 1612 | 85 | |
GenBank mRNA | BC088421.1 | - | 10 - 11 | 1612 | 7123493 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |