Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fus | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012137.1 | NP_001012137.1 | 13 - 14 | 1434 | 80 | |
XM_006230290.2 | XP_006230352.1 | 13 - 14 | 1458 | 80 | ||
GenBank mRNA | BC087153.1 | - | 13 - 14 | 1434 | 7132489 | 80 |
FQ220042.1 | - | 5 - 6 | 543 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |