Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031691.1 | NP_113879.1 | 25 - 26 | 2965 | 105 | |
GenBank mRNA | AF021334.1 | - | 25 - 26 | 2965 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |