Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apobr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109154.1 | NP_001102624.1 | 3 - 4 | 2066 | 68 | |
XM_006230319.2 | XP_006230381.1 | 2 - 3 | 2725 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |