Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032615.2 | NP_116004.2 | 636 | 114 | ||
GenBank mRNA | AF212861.1 | - | 581 | 114 | ||
BC070897.1 | - | 636 | 7099596 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |