Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbrs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001080034.5 | XP_001080034.4 | 7 - 8 | 2755 | 153 | |
XM_006230374.2 | XP_006230436.1 | 7 - 8 | 2755 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |