Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tufm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106295.1 | NP_001099765.1 | 5 - 6 | 1023 | 59 | |
XM_006230224.1 | XP_006230286.1 | 5 - 6 | 1018 | 59 | ||
XM_006230225.1 | XP_006230287.1 | 5 - 6 | 1018 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |