Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmtc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135858.1 | NP_001129330.1 | 6 - 7 | 919 | 91 | |
XM_008765316.1 | XP_008763538.1 | 5 - 6 | 829 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |