Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmbim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199116.2 | NP_954547.1 | 3 - 4 | 378 | 94 | |
GenBank mRNA | BC060596.1 | - | 3 - 4 | 379 | 6889044 | 94 |
FQ217672.1 | - | 3 - 4 | 321 | 94 | ||
FQ228009.1 | - | 3 - 4 | 375 | 94 | ||
FQ233689.1 | - | 3 - 4 | 372 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |