Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241395.2 | XP_006241457.1 | 8 - 9 | 1258 | 106 | |
GenBank mRNA | BC072473.1 | - | 8 - 9 | 1069 | 7100360 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |