Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729781.3 | XP_002729827.2 | 6 - 7 | 747 | 91 | |
XM_008765341.1 | XP_008763563.1 | 6 - 7 | 912 | 91 | ||
XM_008765342.1 | XP_008763564.1 | 5 - 6 | 979 | 91 | ||
GenBank mRNA | FQ231893.1 | - | 6 - 7 | 811 | 91 | |
FQ235303.1 | - | 6 - 7 | 811 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |