Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729781.3 | XP_002729827.2 | 9 - 10 | 1112 | 79 | |
XM_008765341.1 | XP_008763563.1 | 9 - 10 | 1277 | 79 | ||
XM_008765342.1 | XP_008763564.1 | 8 - 9 | 1344 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |