Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sycp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013041.1 | NP_037173.1 | 2 - 3 | 256 | 136 | |
GenBank mRNA | X75785.1 | - | 2 - 3 | 256 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |