Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139216.1 | NP_631962.1 | 4 - 5 | 1946 | 158 | |
GenBank mRNA | M34052.1 | - | 4 - 5 | 1946 | 158 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel 3' Most Rattus norvegicus 1700 Probe spans exons |