Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5dc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134887.1 | NP_001128359.1 | 3 - 4 | 563 | 154 | |
XM_006241203.2 | XP_006241265.1 | 3 - 4 | 564 | 154 | ||
XM_006241204.2 | XP_006241266.1 | 3 - 4 | 565 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |