Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270788.1 | NP_001257717.1 | 7 - 8 | 1017 | 124 | |
NM_139100.2 | NP_620800.2 | 7 - 8 | 1014 | 124 | ||
GenBank mRNA | BC070918.1 | - | 7 - 8 | 956 | 7104964 | 124 |
FQ213208.1 | - | 2 - 3 | 180 | 124 | ||
FQ213570.1 | - | 2 - 3 | 180 | 124 | ||
FQ213750.1 | - | 7 - 8 | 973 | 124 | ||
FQ214401.1 | - | 2 - 3 | 180 | 124 | ||
FQ214581.1 | - | 2 - 3 | 180 | 124 | ||
FQ214698.1 | - | 7 - 8 | 971 | 124 | ||
FQ215697.1 | - | 7 - 8 | 929 | 124 | ||
FQ215957.1 | - | 7 - 8 | 973 | 124 | ||
FQ216349.1 | - | 7 - 8 | 970 | 124 | ||
FQ216361.1 | - | 7 - 8 | 971 | 124 | ||
FQ216421.1 | - | 7 - 8 | 971 | 124 | ||
FQ216474.1 | - | 7 - 8 | 979 | 124 | ||
FQ216716.1 | - | 2 - 3 | 180 | 124 | ||
FQ216978.1 | - | 2 - 3 | 180 | 124 | ||
FQ217343.1 | - | 2 - 3 | 180 | 124 | ||
FQ217707.1 | - | 7 - 8 | 971 | 124 | ||
FQ217883.1 | - | 2 - 3 | 198 | 124 | ||
FQ220537.1 | - | 7 - 8 | 968 | 124 | ||
FQ220626.1 | - | 7 - 8 | 955 | 124 | ||
FQ222611.1 | - | 2 - 3 | 180 | 124 | ||
FQ223916.1 | - | 7 - 8 | 971 | 124 | ||
FQ224211.1 | - | 2 - 3 | 180 | 124 | ||
FQ227446.1 | - | 7 - 8 | 124 | |||
FQ229113.1 | - | 7 - 8 | 968 | 124 | ||
FQ230627.1 | - | 7 - 8 | 970 | 124 | ||
FQ232293.1 | - | 5 - 6 | 773 | 124 | ||
FQ232835.1 | - | 7 - 8 | 964 | 124 | ||
FQ234854.1 | - | 7 - 8 | 968 | 124 | ||
M23984.1 | - | 6 - 7 | 933 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |