Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Helb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241426.2 | XP_006241488.1 | 6 - 7 | 2335 | 125 | |
XM_006241428.2 | XP_006241490.1 | 5 - 6 | 2088 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |