Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Best3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191783.1 | NP_001178712.1 | 4 - 5 | 631 | 90 | |
XM_008765394.1 | XP_008763616.1 | 6 - 7 | 1775 | 90 | ||
XM_008765395.1 | XP_008763617.1 | 4 - 5 | 1512 | 90 | ||
GenBank mRNA | FQ222375.1 | - | 6 - 7 | 1126 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |