Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108080.1 | NP_001101550.1 | 4 - 5 | 359 | 92 | |
XM_006241223.2 | XP_006241285.1 | 5 - 6 | 538 | 92 | ||
XM_006241224.2 | XP_006241286.1 | 5 - 6 | 515 | 92 | ||
XM_006241225.2 | XP_006241287.1 | 4 - 5 | 442 | 92 | ||
XM_006241226.2 | XP_006241288.1 | 4 - 5 | 418 | 92 | ||
GenBank mRNA | FQ233971.1 | - | 5 - 6 | 469 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |