Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC682526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008758875.1 | XP_008757097.1 | 92 |
Gene Symbol: Vom2r17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099491.1 | NP_001092961.1 | 4 - 5 | 92 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Rattus norvegicus Probe spans exons |