Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sephs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079889.2 | NP_001073358.2 | 1 | 2098 | 61 | |
GenBank mRNA | BC081915.1 | - | 1 | 1993 | 61 | |
BC097429.1 | - | 1 | 2098 | 61 | ||
BC128783.1 | - | 1 | 1955 | 7378558 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon |