Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009455.1 | NP_001009455.1 | 5 - 6 | 2198 | 50 | |
GenBank mRNA | BC087086.1 | - | 5 - 6 | 2198 | 7189809 | 50 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |