Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080401.1 | NP_536326.1 | 1 - 2 | 332 | 88 | |
XM_006229718.2 | XP_006229780.1 | 1 - 2 | 184 | 88 | ||
XM_006229719.2 | XP_006229781.1 | 2 - 3 | 633 | 88 | ||
XM_006229720.2 | XP_006229782.1 | 2 - 3 | 1555 | 88 | ||
XM_008759667.1 | XP_008757889.1 | 1 - 2 | 69 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |