Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: l7Rn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013897.1 | NP_001013919.1 | 3 - 4 | 510 | 80 | |
XM_006229662.2 | XP_006229724.1 | 2 - 3 | 477 | 80 | ||
GenBank mRNA | BC088340.1 | - | 3 - 4 | 510 | 7367791 | 80 |
FQ212130.1 | - | 2 - 3 | 421 | 80 | ||
FQ213824.1 | - | 2 - 3 | 422 | 80 | ||
FQ214677.1 | - | 2 - 3 | 422 | 80 | ||
FQ215506.1 | - | 2 - 3 | 413 | 80 | ||
FQ216197.1 | - | 2 - 3 | 422 | 80 | ||
FQ216668.1 | - | 2 - 3 | 422 | 80 | ||
FQ216759.1 | - | 2 - 3 | 422 | 80 | ||
FQ218863.1 | - | 2 - 3 | 419 | 80 | ||
FQ220044.1 | - | 2 - 3 | 422 | 80 | ||
FQ220575.1 | - | 2 - 3 | 422 | 80 | ||
FQ224272.1 | - | 2 - 3 | 424 | 80 | ||
FQ229506.1 | - | 2 - 3 | 418 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |