Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_344962.7 | XP_344963.5 | 2 - 3 | 1416 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |