Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exnef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079706.1 | NP_001073174.1 | 8 - 9 | 875 | 128 | |
XM_006230111.2 | XP_006230173.1 | 7 - 8 | 805 | 128 | ||
XM_006230121.2 | XP_006230183.1 | 4 - 5 | 757 | 128 | ||
XM_008759751.1 | XP_008757973.1 | 7 - 8 | 811 | 128 | ||
XM_008759752.1 | XP_008757974.1 | 7 - 8 | 1096 | 128 | ||
XM_008759753.1 | XP_008757975.1 | 7 - 8 | 1097 | 128 | ||
XM_008759754.1 | XP_008757976.1 | 7 - 8 | 811 | 128 | ||
XM_008759755.1 | XP_008757977.1 | 7 - 8 | 1096 | 128 | ||
XM_008759756.1 | XP_008757978.1 | 7 - 8 | 1096 | 128 | ||
XM_008759757.1 | XP_008757979.1 | 5 - 6 | 856 | 128 | ||
XM_008759758.1 | XP_008757980.1 | 7 - 8 | 1098 | 128 | ||
XM_008759759.1 | XP_008757981.1 | 6 - 7 | 584 | 128 | ||
XM_008759760.1 | XP_008757982.1 | 7 - 8 | 811 | 128 | ||
GenBank mRNA | BC128775.1 | - | 8 - 9 | 875 | 7133646 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |